Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Alcohol Hydrometers, Proof Scale, IRS Specification, with Certification

Internal revenue size R, 185...206 proof, ±0.2% tol., 305 mm (12"), 0.2
  76546-528
 :  6613-R-C
Alcohol Hydrometers, Proof Scale, IRS Specification, with Certification
  76546-528
Alcohol Hydrometers, Proof Scale, IRS Specification, with Certification
Alcohol Hydrometers, Proof Scale, IRS Specification, with Certification

Internal revenue size R, 185...206 proof, ±0.2% tol., 305 mm (12"), 0.2

 
$617.19
 
   
Each
 

These IRS alcohol hydrometers use the Proof scale and comply with IRS specifications.

  • Calibration for use at 15.5 °C/60 °F
  • NIST-Traceable certificate

Verify the alcohol content of your ethyl alcohol-based spirits with our proof scale hydrometer. Tolerances match those laid out by IRS specifications.

 : Includes hydrometer, NIST-traceable certificate, and PVC case.

 
  • Désignation:
    Internal revenue size R
  • Longueur:
    305 mm (12")
  • Range:
    185...206 proof, ±0.2% tol.
  • Subdivisions:
    0.2