Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Precision Short Form Specific Gravity Hydrometers

Precision Short Form Specific Gravity Hydrometers

1.060...1.130, 165 mm (6.5"), 0.001
Catalog # 76546-362
Supplier:   Heathrow Scientific
Supplier #:  6602-8S
Precision Short Form Specific Gravity Hydrometers
Catalog # 76546-362
Precision Short Form Specific Gravity Hydrometers
Precision Short Form Specific Gravity Hydrometers

1.060...1.130, 165 mm (6.5"), 0.001

Price
$48.91
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents

These precision short form specific gravity hydrometers use less volume of the test liquid while still offering 0.001 resolution.

Each of these hydrometers has a small specific gravity range and has been calibrated for use at 60 °F.

Spécifications
  • Longueur:
    165 mm (6.5")
  • Range:
    1.060...1.130
  • Subdivisions:
    0.001
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.