Precision Short Form Specific Gravity Hydrometers
0.880...0.950, 165 mm (6.5"), 0.001
  76546-350
 :  6602-5S
Precision Short Form Specific Gravity Hydrometers
  76546-350
Precision Short Form Specific Gravity Hydrometers
Precision Short Form Specific Gravity Hydrometers
0.880...0.950, 165 mm (6.5"), 0.001
$48.91
Each
These precision short form specific gravity hydrometers use less volume of the test liquid while still offering 0.001 resolution.
Each of these hydrometers has a small specific gravity range and has been calibrated for use at 60 °F.
  • Longueur:
    165 mm (6.5")
  • Range:
    0.880...0.950
  • Subdivisions:
    0.001