Precision Short Form Specific Gravity Hydrometers
0.820...0.890, 165 mm (6.5"), 0.001
Catalog # 76546-346
Supplier: Heathrow Scientific
Fournisseur #: 6602-4S
Precision Short Form Specific Gravity Hydrometers
Catalog # 76546-346
Supplier: Heathrow Scientific
Jump to:
Product Details & Documents
These precision short form specific gravity hydrometers use less volume of the test liquid while still offering 0.001 resolution.
Each of these hydrometers has a small specific gravity range and has been calibrated for use at 60 °F.
Spécifications
- Longueur:165 mm (6.5")
- Range:0.820...0.890
- Subdivisions:0.001
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



