Precision Short Form Specific Gravity Hydrometers
1.180...1.250, 165 mm (6.5"), 0.001
Catalog # 76546-326
Supplier: Heathrow Scientific
Fournisseur #: 6602-10S
Precision Short Form Specific Gravity Hydrometers
Catalog # 76546-326
Supplier: Heathrow Scientific
Jump to:
Product Details & Documents
These precision short form specific gravity hydrometers use less volume of the test liquid while still offering 0.001 resolution.
Each of these hydrometers has a small specific gravity range and has been calibrated for use at 60 °F.
Spécifications
- Longueur:165 mm (6.5")
- Range:1.180...1.250
- Subdivisions:0.001
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



