Vee Gee Filter Holder Components
Vee Gee Filter Holder Components
PTFE gaskets
Catalog # 76546-264
Supplier:   Heathrow Scientific
Supplier #:  6168-4741
Vee Gee Filter Holder Components
Catalog # 76546-264
Vee Gee Filter Holder Components
Vee Gee Filter Holder Components
PTFE gaskets
Price
$18.43
New Customer Promotional Price
Log in for availability
Pack of 10
Product Details & Documents
Available with fritted glass support base for general and biological applications, or with removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.
  • For use with Vee Gee Filter system kits
All items except the PTFE gasket are autoclavable.
Specifications
  • Désignation:
    PTFE gaskets