Vee Gee Filter Holder Components
PTFE gaskets
Catalog # 76546-264
Supplier: Heathrow Scientific
Supplier #: 6168-4741
Jump to:
Product Details & Documents
Available with fritted glass support base for general and biological applications, or with removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.
- For use with Vee Gee Filter system kits
All items except the PTFE gasket are autoclavable.
Specifications
- Désignation:PTFE gaskets