Vee Gee Filter Holder Components
Stainless steel screen
  76546-262
 :  6168-4731
Vee Gee Filter Holder Components
  76546-262
Vee Gee Filter Holder Components
Vee Gee Filter Holder Components
Stainless steel screen
$78.60
Each
Available with fritted glass support base for general and biological applications, or with removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.
  • For use with Vee Gee Filter system kits
All items except the PTFE gasket are autoclavable.
  • Désignation:
    Stainless steel screen
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.