Vee Gee Filter Holder Components
Vee Gee Filter Holder Components
Glass support base, fritted
Catalog # 76546-260
Supplier:   Heathrow Scientific
Supplier #:  6168-4722
Vee Gee Filter Holder Components
Catalog # 76546-260
Vee Gee Filter Holder Components
Vee Gee Filter Holder Components
Glass support base, fritted
Price
$81.32
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Available with fritted glass support base for general and biological applications, or with removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.
  • For use with Vee Gee Filter system kits
All items except the PTFE gasket are autoclavable.
Spécifications
  • Désignation:
    Glass support base, fritted