Vee Gee Filter Holder Components
Glass support base, for stainless steel screen
Catalog # 76546-258
Supplier: Heathrow Scientific
Fournisseur #: 6168-4721
Jump to:
Product Details & Documents
Available with fritted glass support base for general and biological applications, or with removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.
- For use with Vee Gee Filter system kits
All items except the PTFE gasket are autoclavable.
Spécifications
- Désignation:Glass support base, for stainless steel screen