Vee Gee Filter Holder Components
Funnel, 100 ml
Catalog # 76546-254
Supplier:   Heathrow Scientific
 :  6168-47100
Vee Gee Filter Holder Components
Catalog # 76546-254
Vee Gee Filter Holder Components
Vee Gee Filter Holder Components
Funnel, 100 ml
Price
102,16 $
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Available with fritted glass support base for general and biological applications, or with removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.
  • For use with Vee Gee Filter system kits
All items except the PTFE gasket are autoclavable.
Specifications
  • Désignation:
    Funnel, 100 ml