Vee Gee Filter Holder Components
Funnel, 100 ml
Catalog # 76546-254
Supplier: Heathrow Scientific
: 6168-47100
Jump to:
Product Details & Documents
Available with fritted glass support base for general and biological applications, or with removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.
- For use with Vee Gee Filter system kits
All items except the PTFE gasket are autoclavable.
Specifications
- Désignation:Funnel, 100 ml