Vee Gee Filter Holder Systems
Stainless steel support base, Ultraclean filtrate and proteinaceous solutions, 100 ml
  76546-252
 :  6168-0004
Vee Gee Filter Holder Systems
  76546-252
Vee Gee Filter Holder Systems
Vee Gee Filter Holder Systems
Stainless steel support base, Ultraclean filtrate and proteinaceous solutions, 100 ml
305,94 $
Each
Filtration filter holders are offered with a wide variety of components for different applications.
  • Available in 100 or 300 ml capacities
Available with a fritted glass support base for general and biological applications, or with a removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.
  • Désignation:
    Stainless steel support base
  • For Use With:
    Ultraclean filtrate and proteinaceous solutions
  • Funnel Capacity:
    100 ml