Vee Gee Filter Holder Systems
Fritted glass support base, General and biological applications, 100 ml
  76546-250
 :  6168-0003
Vee Gee Filter Holder Systems
  76546-250
Vee Gee Filter Holder Systems
Vee Gee Filter Holder Systems
Fritted glass support base, General and biological applications, 100 ml
$263.47
Each
Filtration filter holders are offered with a wide variety of components for different applications.
  • Available in 100 or 300 ml capacities
Available with a fritted glass support base for general and biological applications, or with a removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.
  • Désignation:
    Fritted glass support base
  • For Use With:
    General and biological applications
  • Funnel Capacity:
    100 ml