Vee Gee Filter Holder Systems
Stainless steel support base, Ultraclean filtrate and proteinaceous solutions, 300 ml
Catalog # 76546-248
Supplier: Heathrow Scientific
Supplier #: 6168-0002
Jump to:
Product Details & Documents
Filtration filter holders are offered with a wide variety of components for different applications.
- Available in 100 or 300 ml capacities
Available with a fritted glass support base for general and biological applications, or with a removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.
Specifications
- Désignation:Stainless steel support base
- For Use With:Ultraclean filtrate and proteinaceous solutions
- Funnel Capacity:300 ml