Vee Gee Filter Holder Systems
Vee Gee Filter Holder Systems
Stainless steel support base, Ultraclean filtrate and proteinaceous solutions, 300 ml
Catalog # 76546-248
Supplier:   Heathrow Scientific
Supplier #:  6168-0002
Vee Gee Filter Holder Systems
Catalog # 76546-248
Vee Gee Filter Holder Systems
Vee Gee Filter Holder Systems
Stainless steel support base, Ultraclean filtrate and proteinaceous solutions, 300 ml
Price
345,16 $
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Filtration filter holders are offered with a wide variety of components for different applications.
  • Available in 100 or 300 ml capacities
Available with a fritted glass support base for general and biological applications, or with a removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.
Specifications
  • Désignation:
    Stainless steel support base
  • For Use With:
    Ultraclean filtrate and proteinaceous solutions
  • Funnel Capacity:
    300 ml