Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Vee Gee Micro Volumetric Flasks | Avantor

Vee Gee Micro Volumetric Flasks

2 ml, #9, ±0.015 ml
Catalog # 76546-134
Supplier:   Heathrow Scientific
Fournisseur #:  2306A-2
Vee Gee Micro Volumetric Flasks
Catalog # 76546-134
Vee Gee Micro Volumetric Flasks
Vee Gee Micro Volumetric Flasks

2 ml, #9, ±0.015 ml

Price
$158.54
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 6 Case of 576
Product Details & Documents

These micro volumetric flasks feature machine-blown bodies and heavy-beaded, heavy-tubing necks that are tooled for standard taper glass stoppers.

  • Made of premium borosilicate glass with a low coefficient of expansion
  • Designed 'to contain' with a temperature of calibration at 20 °C (68 °F)
  • The graduation line is sharply defined and of uniform width
  • Each flask has a large marking square and includes a glass stopper
  • Features a hexagonal base for added stability

Our flasks are constructed of premium borosilicate glass, ensuring exceptional clarity and resistance to chemical interactions. Each volumetric flask is supplied with a standard taper glass stopper which provides a leakproof seal. This airtight seal not only preserves the integrity of your samples but also enhances the reliability of your volumetric measurements.

Certifications: Meets ASTM E-288 Class A specifications and USP standards for glassware.

Spécifications
  • Capacité:
    2 ml
  • ST Stopper No.:
    #9
  • Tolerance:
    ±0.015 ml
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.