Smart Gasket G-TH-150-S-4 1 1/2" Tri-Clamp, 4 Port, Silicone, Each
Catalogue # MFLX30545-12
Fournisseur: VWR International
À propos de cet article
Smart Gasket lets you take samples or readings from a sanitary line.
- Measure temperature, take a sample, or dose chemicals into your process line
- Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
- Sanitary clamp size (Port 1):1½"
- Connection type:Sanitary
- Description:1 1/2" Tri-Clamp, 4 Port, Silicone, Each
- Environmentally Preferable:
- No. of ports:4
- Warranty:1 year
- Cat. no.:MFLX30545-12
- Fitting Material:Platinum-cured silicone
- Sanitary Gasket Size:1½"
Frequently Bought Together