BASIC* Lab Coats, Halyard | Avantor
BASIC* Lab Coats, Halyard
White, XL
Catalog # CA76540-358
Fournisseur #:  10123
BASIC* Lab Coats, Halyard
Catalog # CA76540-358
BASIC* Lab Coats, Halyard
BASIC* Lab Coats, Halyard
White, XL
Price
$232.98
Log in pour la disponibilité
Case of 25
Product Details & Documents
HALYARD* BASIC* Lab Coats all feature a protective 3-layer fabric that offers exceptional resistance to blood strike-through at various levels of exposure.
  • Traditional collar
  • Raglan sleeves
  • Knit cuffs
  • Four snap front closure
  • Two reinforced pockets
  • Size/name label
Spécifications
  • Taille de confection:
    XL
  • Color:
    White