BASIC* Lab Coats, Halyard
White, Small
Catalog # CA76540-352
Supplier: O&M HALYARD CANADA, INC CA
: 10120
Jump to:
Product Details & Documents
HALYARD* BASIC* Lab Coats all feature a protective 3-layer fabric that offers exceptional resistance to blood strike-through at various levels of exposure.
- Traditional collar
- Raglan sleeves
- Knit cuffs
- Four snap front closure
- Two reinforced pockets
- Size/name label
Specifications
- Taille de confection:Small
- Color:White