BASIC PLUS* Lab Coats, Halyard
White, Small
Catalog # CA76540-342
Supplier: O&M HALYARD CANADA, INC CA
Supplier #: 10020-66
Jump to:
Product Details & Documents
HALYARD* BASIC PLUS* Lab Coats all feature a protective 3-layer fabric that offers exceptional resistance to blood strike-through at various levels of exposure.
- Knit collar and cuffs
- Five snap front closure
- Three reinforced pockets
- Neck loop for hanging
- Size/name label
Spécifications
- Taille de confection:Small
- Color:White