BASIC PLUS* Lab Coats, Halyard
Blue, Large
  CA76540-336
 :  10032-66
BASIC PLUS* Lab Coats, Halyard
  CA76540-336
BASIC PLUS* Lab Coats, Halyard
BASIC PLUS* Lab Coats, Halyard
Blue, Large
$250.62
Case of 25
HALYARD* BASIC PLUS* Lab Coats all feature a protective 3-layer fabric that offers exceptional resistance to blood strike-through at various levels of exposure.
  • Knit collar and cuffs
  • Five snap front closure
  • Three reinforced pockets
  • Neck loop for hanging
  • Size/name label
  • Taille de confection:
    Large
  • Color:
    Blue