BASIC PLUS* Lab Coats, Halyard
Blue, Medium
Catalog # CA76540-334
 :  10031-66
BASIC PLUS* Lab Coats, Halyard
Catalog # CA76540-334
BASIC PLUS* Lab Coats, Halyard
BASIC PLUS* Lab Coats, Halyard
Blue, Medium
Price
207,54 $
Log in pour la disponibilité
Case of 25
Product Details & Documents
HALYARD* BASIC PLUS* Lab Coats all feature a protective 3-layer fabric that offers exceptional resistance to blood strike-through at various levels of exposure.
  • Knit collar and cuffs
  • Five snap front closure
  • Three reinforced pockets
  • Neck loop for hanging
  • Size/name label
Spécifications
  • Taille de confection:
    Medium
  • Color:
    Blue