Powder Scoops, Polypropylene | Avantor
Powder Scoops, Polypropylene
Powder scoop, PP, 5 ml
Catalog # 470337-868
Fournisseur #:  UNSC005
Powder Scoops, Polypropylene
Catalog # 470337-868
Powder Scoops, Polypropylene
Powder Scoops, Polypropylene
Powder scoop, PP, 5 ml
Price
$62.25
Log in pour la disponibilité
Pack of 100
Product Details & Documents
Convenient scoops for measuring out powders.
  • Polypropylene
  • Two sizes available
  • Do not microwave
These versatile scoops are ideal for measuring powders or liquids.
Spécifications
  • Capacité:
    5 ml
  • Désignation:
    Powder scoop
  • Matière:
    PP