Powder Scoops, Polypropylene
Powder scoop, PP, 5 ml
Catalog # 470337-868
Supplier: United Scientific Supplies
Fournisseur #: UNSC005
Jump to:
Product Details & Documents
Convenient scoops for measuring out powders.
- Polypropylene
- Two sizes available
- Do not microwave
These versatile scoops are ideal for measuring powders or liquids.
Spécifications
- Capacité:5 ml
- Désignation:Powder scoop
- Matière:PP