Powder Scoops, Polypropylene | Avantor
Powder Scoops, Polypropylene
Powder scoop, PP, 1 ml
Catalog # 470337-866
Fournisseur #:  UNSC001
Powder Scoops, Polypropylene
Catalog # 470337-866
Powder Scoops, Polypropylene
Powder Scoops, Polypropylene
Powder scoop, PP, 1 ml
Price
$56.40
Log in pour la disponibilité
Pack of 100
Product Details & Documents
Convenient scoops for measuring out powders.
  • Polypropylene
  • Two sizes available
  • Do not microwave
These versatile scoops are ideal for measuring powders or liquids.
Spécifications
  • Capacité:
    1 ml
  • Désignation:
    Powder scoop
  • Matière:
    PP