Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Mag-Bind® Universal Pathogen Core Kit | Avantor

Mag-Bind® Universal Pathogen Core Kit

Universal pathogen kit, 4×96 preps
Catalog # CA76539-254
Supplier:   Omega Bio-Tek
Fournisseur #:  M4030-01
Mag-Bind® Universal Pathogen Core Kit
Catalog # CA76539-254
Supplier:  Omega Bio-Tek
CAS Number:  
Mag-Bind® Universal Pathogen Core Kit

Universal pathogen kit, 4×96 preps

CAS Number:  
Price
$2,054.85
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Isolate host genomic, bacterial, fungal spore, and viral DNA and viral RNA from tissue, urine, serum, and fecal samples using magnetic beads.

  • Purified DNA is suitable for PCR, restriction digestion, and hybridization applications

The Mag-Bind® Universal Pathogen Core Kit allows rapid and reliable isolation of high-quality host genomic DNA, gram-positive and negative bacterial DNA, fungal spore DNA, viral DNA, and viral RNA from tissue, urine, serum, and fecal samples. The system incorporates a recommended enzymatic lysis which allows for fully automated extraction post-lysis via Hamilton STAR™, Thermo KingFisher™ Flex, Applied Biosystems® MagMAX™, Qiagen Biosprint, Tecan Fluent® Automation Workstation, Tecan Freedom EVO®, and other liquid handling instruments. There are no organic extractions, thus reducing plastic waste and hands-on time and multiple samples can be processed in parallel. The system combines the Mag-Bind® technology with RBB Buffer to eliminate PCR inhibiting compounds within the samples. Purified DNA is suitable for PCR, restriction digestion, and hybridization applications.

Spécifications

Spécifications

  • Désignation:
    Universal pathogen kit
  • Taille:
    4×96 preps
  • Cat. No.:
    CA76539-254
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Magnetic beads
  • Sample type:
    Bacteria

Spécifications