Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Mag-Bind® Universal Pathogen Core Kit

Mag-Bind® Universal Pathogen Core Kit

Universal pathogen kit, 1×96 preps
Catalog # CA76539-252
Supplier:   Omega Bio-Tek
Supplier #:  M4030-00
Mag-Bind® Universal Pathogen Core Kit
Catalog # CA76539-252
Supplier:  Omega Bio-Tek
CAS Number:  
Mag-Bind® Universal Pathogen Core Kit

Universal pathogen kit, 1×96 preps

CAS Number:  
Price
$590.77
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Isolate host genomic, bacterial, fungal spore, and viral DNA and viral RNA from tissue, urine, serum, and fecal samples using magnetic beads.

  • Purified DNA is suitable for PCR, restriction digestion, and hybridization applications

The Mag-Bind® Universal Pathogen Core Kit allows rapid and reliable isolation of high-quality host genomic DNA, gram-positive and negative bacterial DNA, fungal spore DNA, viral DNA, and viral RNA from tissue, urine, serum, and fecal samples. The system incorporates a recommended enzymatic lysis which allows for fully automated extraction post-lysis via Hamilton STAR™, Thermo KingFisher™ Flex, Applied Biosystems® MagMAX™, Qiagen Biosprint, Tecan Fluent® Automation Workstation, Tecan Freedom EVO®, and other liquid handling instruments. There are no organic extractions, thus reducing plastic waste and hands-on time and multiple samples can be processed in parallel. The system combines the Mag-Bind® technology with RBB Buffer to eliminate PCR inhibiting compounds within the samples. Purified DNA is suitable for PCR, restriction digestion, and hybridization applications.

Specifications

Specifications

  • Désignation:
    Universal pathogen kit
  • Taille:
    1×96 preps
  • Cat. No.:
    CA76539-252
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Magnetic beads
  • Sample type:
    Bacteria

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.