VWR® Wall Shelf Brackets, Fixed and Adjustable
Vertical Wall Support, Snow white, Adjustable shelf Brackets (Max. 3 shelves), 48"
Catalog # CWF-0170-04
Supplier: VWR International
Jump to:
Product Details & Documents
Fixed and adjustable wall-mounted brackets.
- Designed to suit a wide array of storage applications
- Fixed brackets mount directly to the wall
- Adjustable brackets are designed to mount to vertical wall supports (sold separately)
Fixed brackets features a flat top surface for simple shelf attachment. Adjustable brackets features a lipped top surface to provide increased security and stability when shelves are attached.
Ordering information: Fixed shelf brackets are supplied without mounting hardware. Adjustable brackets come with mounting hardware.
Spécifications
- Color:Snow white
- Désignation:Vertical Wall Support
- Hauteur:48"
- For Use With:Adjustable shelf Brackets (Max. 3 shelves)



