Styrofoam Head Models | Avantor
Styrofoam Head Models
Female, 6.3×6.8×10"
Catalog # 470337-664
Fournisseur #:  RS260/6
Styrofoam Head Models
Catalog # 470337-664
Styrofoam Head Models
Styrofoam Head Models
Female, 6.3×6.8×10"
Price
$23.60
Log in pour la disponibilité
Each Case of 6
Product Details & Documents
Styrofoam head models for multiple uses.
  • Available in male, female, and faceless versions
  • Smooth surface paints easily
  • Dense and durable
These smooth foam head forms can be painted or finished, allowing students to make or display their own models. Use them as a base to create brain maps, or sculpt masks. The foam is denser than most styrofoam, so it can also be trimmed or cut to create internal anatomical models. They may also be used for basic modeling projects in multiple disciplines.
Spécifications
  • Désignation:
    Female
  • Taille:
    6.3×6.8×10"