Translucent Snap Caps, Globe Scientific
13 mm, Yellow
76538-876
: 113143Y
These colorful snap-on closures are an effective way to recap blood drawing tubes, glass test tubes and plastic culture tubes.
- Economical way to recap glass tubes, regardless of manufacturer
- Available in seven (7) translucent colors that correspond to Vacutainer® cap colors
- Caps can be written on with indelible ink for easy sample identification
- Prevents spills and evaporation by employing a liquid tight seal
These caps are precision molded from low-density polyethylene (LDPE) and maintain a tight seal during centrifugation, agitation and refrigeration procedures. They are available in bright translucent colors for sample identification purposes. The caps feature a single thumb tab for easy removal and close nesting in tube racks.
- Color:Yellow
- For Tube O.D.:13 mm