Translucent Snap Caps, Globe Scientific
Translucent Snap Caps, Globe Scientific
16 mm, Blue
Catalog # 76538-868
Supplier:   Globe Scientific
Supplier #:  113144B
Translucent Snap Caps, Globe Scientific
Catalog # 76538-868
Supplier:  Globe Scientific
Translucent Snap Caps, Globe Scientific
Translucent Snap Caps, Globe Scientific
16 mm, Blue
Price
$66.88
New Customer Promotional Price
Log in for availability
Pack of 1,000
Product Details & Documents
These colorful snap-on closures are an effective way to recap blood drawing tubes, glass test tubes and plastic culture tubes.
  • Economical way to recap glass tubes, regardless of manufacturer
  • Available in seven (7) translucent colors that correspond to Vacutainer® cap colors
  • Caps can be written on with indelible ink for easy sample identification
  • Prevents spills and evaporation by employing a liquid tight seal
These caps are precision molded from low-density polyethylene (LDPE) and maintain a tight seal during centrifugation, agitation and refrigeration procedures. They are available in bright translucent colors for sample identification purposes. The caps feature a single thumb tab for easy removal and close nesting in tube racks.
Specifications
  • Color:
    Blue
  • For Tube O.D.:
    16 mm