Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
SevenDirect™ SD20 pH Meter | Avantor

SevenDirect™ SD20 pH Meter

pH SevenDirect™ SD20 organic kit
Catalog # 76533-440
Fournisseur #:  30671557
SevenDirect™ SD20 pH Meter
Catalog # 76533-440
Supplier:  
SevenDirect™ SD20 pH Meter
SevenDirect™ SD20 pH Meter

pH SevenDirect™ SD20 organic kit

Discontinued
Product Details & Documents

The pH meter SevenDirect™ SD20 provides accurate pH measurements for almost any sample across a wide range of applications. The EasyPlace™ electrode arm and protective cover are included with the instrument. The benchtop meter's ease of use, data handling options and robustness make it ideal for all industries.

  • Intuitive user interface using 7" touchscreen
  • On-screen instructions guide through the calibration process
  • EasyPlace™ electrode arm ensures precise placement of the sensor
  • Feature a closed connector compartment to prevent corrosion
  • A replacable protective cover keeps the instrument safe from dust and spills (IP 54 in-use)
  • Instruments support GLP compliance by storing key measurement data

The intuitive user interface of the 7" touchscreen facilitates the measurement process by avoiding unnecessary steps and focusing on the most important information. The SevenDirect™ has a guided verification process that provides a straightforward way to check system performance with a clear pass/fail indication. The meter has a replaceable protective cover to keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use).

Delivery information: The meter-only model includes EasyPlace™ electrode holder, protective cover, Get-Started guide and power supply. The kits also include an InLab sensor and 2 buffer sachets for pH 4.01, 7.00, 9.21 and 10.00, in addition to the meter-only standard accessories.

Spécifications
  • Désignation:
    pH SevenDirect™ SD20 organic kit
  • Cat. No.:
    76533-440
  • pH accuracy:
    0.002
  • pH resolution:
    0.1; 0.01; 0.001
  • Temperature range:
    –30...130 °C
  • mV accuracy:
    0.1
  • mV resolution:
    1; 0.1
Accessories

Informations sur le produit

  • Catalog #
    Details
    Availability
    Price
  • Parking beaker EasyPlace™
    Parking beaker EasyPlace™
    76533-476
    The parking beaker EasyPlace™ for the SevenDirect™ pH meter is robust and resistant to chemicals for most common lab applications. The beaker has convenient 20 mL graduations for quick and easy measurements.  
    Log in pour la disponibilité
    Each - $33.90

Informations sur le produit

  • Catalog #
    Details
    Availability
    Price
  • Parking beaker insert
    Parking beaker insert
    76533-478
    The parking beaker insert EasyPlace™ for the SevenDirect™ pH meter is secure, keeping the beaker in place while working with various lab samples.  
    Log in pour la disponibilité
    Each - $13.20

Informations sur le produit

  • Catalog #
    Details
    Availability
    Price
  • Sachet holder EasyPlace™
    Sachet holder EasyPlace™
    76533-480
    The sachet holder EasyPlace™ for the SevenDirect™ pH meter is extremely convenient for holding sachets in place. The EasyPlace™ sachet holder is made for the newly designed pH and conductivity style sachets to ensure calibration is quick and easy.  
    Log in pour la disponibilité
    Each - $69.30

Informations sur le produit

  • Catalog #
    Details
    Availability
    Price
  • Sensor holder EasyPlace™ triple
    Sensor holder EasyPlace™ triple
    76533-484
    The sensor holder EasyPlace™ for the SevenDirect™ pH meter is extremely convenient for holding sensors in place to ensure accurate measurements. 
    Log in pour la disponibilité
    Each - $41.60

Informations sur le produit

  • Catalog #
    Details
    Availability
    Price
  • Pole EasyPlace™ 30 cm
    Pole EasyPlace™ 30 cm
    76533-488
    The pole EasyPlace™ for the SevenDirect™ pH meter is attached to the base plate to hold electrodes securely and safely. 
    Log in pour la disponibilité
    Each - $339.00

Informations sur le produit

  • Catalog #
    Details
    Availability
    Price
  • Pole EasyPlace™ 50 cm
    Pole EasyPlace™ 50 cm
    76533-490
    The pole EasyPlace™ for the SevenDirect™ pH meter is attached to the base plate to hold electrodes securely and safely. 
    Log in pour la disponibilité
    Each - $402.00

Informations sur le produit

  • Catalog #
    Details
    Availability
    Price
  • Base plate EasyPlace™
    Base plate EasyPlace™
    76533-492
    The base plate EasyPlace™ for the SevenDirect™ meter is extremely secure, distributing weight evenly over the entire meter area. Meter parts such as the electrode arm will be attached to the base plate when you start to build your SevenDirect pH meter.  
    Log in pour la disponibilité
    Each - $203.00

Informations sur le produit

  • Catalog #
    Details
    Availability
    Price
  • Power supply 18W
    Power supply 18W
    76533-494
    The power supply (18 W) can be used as an external power source for the SevenDirect™ pH meter. 
    Log in pour la disponibilité
    Each - $135.00

Informations sur le produit

  • Catalog #
    Details
    Availability
    Price
  • Plug set 18 W power supply
    Plug set 18 W power supply
    76533-496
    The plug set power supply (18 W) can be used as an external power source for the SevenDirect™ pH meter. Additional adaptors included in the plug set. 
    Log in pour la disponibilité
    Each - $45.70

Informations sur le produit

  • Catalog #
    Details
    Availability
    Price
  • Top cover pole EasyPlace™
    Top cover pole EasyPlace™
    76533-498
    The top cover pole EasyPlace™ fits tightly over the EasyPlace™ stand for an added layer of protection when not operating the meter. 
    Log in pour la disponibilité
    Each - $33.90

Informations sur le produit

  • Catalog #
    Details
    Availability
    Price
  • Screws and feet set EasyPlace™
    Screws and feet set EasyPlace™
    76533-500
    The EasyPlace™ screws and feet set are included with the base plate for SevenDirect™ meter assembly. 
    Log in pour la disponibilité
    Each - $41.60

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.