Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
1-1/2" Tri-Clamp, 1 Port, Silicone; 1/EA, 1½", Platinum-Cured Silicone, 1½"
Supplier: VWR International
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Catalog # MFLX30542-12
Supplier: VWR International
Jump to:
Product Details & Documents
Smart Gasket lets you take samples or readings from a sanitary line.
- Measure temperature, take a sample, or dose chemicals into your process line
- Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
Spécifications
- Sanitary clamp size (Port 1):1½"
- Désignation:1-1/2" Tri-Clamp, 1 Port, Silicone; 1/EA
- Fitting Material:Platinum-Cured Silicone
- Sanitary Gasket Size:1½"
- Cat. No.:MFLX30542-12
- Warranty:1 year
- Connection type:Sanitary
- No. of ports:1