Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Sanitary Sampling Gasket, Silicone, 1 Port, 1/2" Tri-Clamp; 1/EA, ¾", Platinum-Cured Silicone, ½"
Catalog # MFLX30542-00
Supplier: VWR International
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Catalog # MFLX30542-00
Supplier: VWR International
Jump to:
Product Details & Documents
Smart Gasket lets you take samples or readings from a sanitary line.
- Measure temperature, take a sample, or dose chemicals into your process line
- Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
Spécifications
- Sanitary clamp size (Port 1):¾"
- Désignation:Sanitary Sampling Gasket, Silicone, 1 Port, 1/2" Tri-Clamp; 1/EA
- Fitting Material:Platinum-Cured Silicone
- Sanitary Gasket Size:½"
- Cat. No.:MFLX30542-00
- Warranty:1 year
- Connection type:Sanitary
- No. of ports:1