Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
L/S® Easy-Load® II Pump Head Mounting Hardware, Four Pump Heads, Stainless Steel. 2 years warranty, Stainless steel, Masterflex L/S® Easy-Load® II Pump head, L/S
Catalog # MFLX77200-04
Supplier: Avantor Fluid Handling
Fournisseur #: 77200-04
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Catalog # MFLX77200-04
Supplier: Avantor Fluid Handling
Jump to:
Product Details & Documents
Easy stack multiple pump heads on your Masterflex® L/S® drive.
- Constructed of durable stainless steel
- See drive specifications for recommended number of heads
Stack multiple pump heads or replacement single pump head mounting hardware. Choose mounting hardware based on the Masterflex® L/S® pump head that you have. Compatible with the Masterflex L/S series pumps.
Spécifications
- Désignation:L/S® Easy-Load® II Pump Head Mounting Hardware, Four Pump Heads, Stainless Steel. 2 years warranty
- Matière:Stainless steel
- Compatible with:Masterflex L/S® Easy-Load® II Pump head
- Pump Series:L/S