Masterflex® Single-Use Forceps, Avantor®
Le produit 76669-824 est une alternative fonctionnelle à MFLX75955-13 mais l'emballage, la quantité, le fabricant et/ou les spécifications peuvent varier.
Product Details & Documents
No more cross-contamination.
- Protect yourself and your samples with disposable forceps
- Autoclavable plastic or stainless steel construction
These single-use forceps are bulk-packed for convenience and affordability. Slide forceps feature an extra-wide grip for greater stability and security when staining cells and tissue. Fine-tipped forceps are ideally suited for work with smaller specimens. Stainless steel forceps have grooved finger grips and ridged tips, best for larger, soft or wet specimens.
Spécifications
- Désignation:Sterile Forceps, stainless steel, 4,5" long; 25/pk
- Matière:Stainless steel
- Overall Length:114 mm (4.5")
- Sterile:+
Accessories
Informations sur le produit
- Catalog #MaterialSterileAvailabilityPrice
- Adhesive coated microscope slides, Poly-l-lysine