PendoTECH® Single-Use Pressure Sensor Accessories
Cable with RJ12 to Midgee Monitor. 6 ft (1.8 m)
MFLX19406-86
CE Compliant
Optimize performance of your PendoTECH® Pressure Sensor.
- Choose from a wide range of replacement parts and accessories
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
- Désignation:Cable with RJ12 to Midgee Monitor. 6 ft (1.8 m)
- Cat. No.:MFLX19406-86
- Warranty:2 years
- Length:182.88 cm (72")