PendoTECH® Single-Use Pressure Sensor Accessories
Adapter Tee, 1/4" Hose Barb x 1/4" Hose Barb x Male Luer Fitting, Polycarbonate
Supplier: PENDOTECH
Fournisseur #: PDKT-103-03
Jump to:
Product Details & Documents
Optimize performance of your PendoTECH® Pressure Sensor.
- Choose from a wide range of replacement parts and accessories
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
Documents
Spécifications
- Désignation:Adapter Tee, 1/4" Hose Barb x 1/4" Hose Barb x Male Luer Fitting, Polycarbonate
- Cat. No.:MFLX19406-34
- Max. pressure:76 psi
- Warranty:3 Months
- Process connection:1/4" x 1/4" hose barb, male luer