PendoTECH® Single-Use Pressure Sensor Accessories
Reusable Temperature Sensor Cable with 1/4" Phone Jack for Luer Sensors; 7 ft (2 m)
Catalog # MFLX19406-14
CE Compliant
Supplier: PENDOTECH
Fournisseur #: PDKT-650-TEMPL
PendoTECH® Single-Use Pressure Sensor Accessories
Catalog # MFLX19406-14
Supplier: PENDOTECH
CE Compliant
Jump to:
Product Details & Documents
Optimize performance of your PendoTECH® Pressure Sensor.
- Choose from a wide range of replacement parts and accessories
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
Documents
Spécifications
- Désignation:Reusable Temperature Sensor Cable with 1/4" Phone Jack for Luer Sensors; 7 ft (2 m)
- Cat. No.:MFLX19406-14
- Warranty:2 years
- Length:213.36 cm (84")