Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
BenchBin™ Benchtop Biohazard Bin | Avantor

BenchBin™ Benchtop Biohazard Bin

Red biohazard bags for BenchBin™
Catalog # 470337-139
Supplier:   MTC BIO, INC.
Fournisseur #:  A9004R
BenchBin™ Benchtop Biohazard Bin
Catalog # 470337-139
Supplier:  MTC BIO, INC.
BenchBin™ Benchtop Biohazard Bin
BenchBin™ Benchtop Biohazard Bin

Red biohazard bags for BenchBin™

Price
$56.05
Log in pour la disponibilité
Each
Product Details & Documents

The BenchBin™ is a compact, convenient, and safe biohazard bin for the benchtop or workstation.

  • Safely dispose of tips, tubes, gloves and more
  • Secures and covers hazardous waste
  • Easy push open/close spring lid
  • Compact footprint (7×5×7")

For the temporary storage of biohazardous material during a lab activity or assay that doesn't require multiple trips to the biohazard bin. The sleek and compact design requires minimal space on the lab bench while the closed-lid system and easy bag removal creates a safe and convenient solution for waste .

Ordering information: Red biohazard bags (8.5×11") are purchased separately in packs of 100.

Delivery information: The BenchBin™ Starter Kit includes one BenchBin and 400 red 8.5×11" autoclave/ biohazard bags.

Spécifications
  • Désignation:
    Red biohazard bags for BenchBin™
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.