Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
BenchBin™ Benchtop Biohazard Bin

BenchBin™ Benchtop Biohazard Bin

BenchBin™ starter kit
Catalog # 470337-138
Supplier:   MTC BIO, INC.
Supplier #:  A8401-SK
BenchBin™ Benchtop Biohazard Bin
Catalog # 470337-138
Supplier:  MTC BIO, INC.
BenchBin™ Benchtop Biohazard Bin
BenchBin™ Benchtop Biohazard Bin

BenchBin™ starter kit

Price
$201.85
Log in for availability
Each
Product Details & Documents

The BenchBin™ is a compact, convenient, and safe biohazard bin for the benchtop or workstation.

  • Safely dispose of tips, tubes, gloves and more
  • Secures and covers hazardous waste
  • Easy push open/close spring lid
  • Compact footprint (7×5×7")

For the temporary storage of biohazardous material during a lab activity or assay that doesn't require multiple trips to the biohazard bin. The sleek and compact design requires minimal space on the lab bench while the closed-lid system and easy bag removal creates a safe and convenient solution for waste .

Ordering information: Red biohazard bags (8.5×11") are purchased separately in packs of 100.

Delivery information: The BenchBin™ Starter Kit includes one BenchBin and 400 red 8.5×11" autoclave/ biohazard bags.

Specifications
  • Désignation:
    BenchBin™ starter kit
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.