Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
BenchBin™ Benchtop Biohazard Bin | Avantor

BenchBin™ Benchtop Biohazard Bin

BenchBin™ benchtop biohazard bin
Catalog # 470337-137
Supplier:   MTC BIO, INC.
Fournisseur #:  A8401B
BenchBin™ Benchtop Biohazard Bin
Catalog # 470337-137
Supplier:  MTC BIO, INC.
BenchBin™ Benchtop Biohazard Bin
BenchBin™ Benchtop Biohazard Bin

BenchBin™ benchtop biohazard bin

Price
$72.35
Log in pour la disponibilité
Each
Product Details & Documents

The BenchBin™ is a compact, convenient, and safe biohazard bin for the benchtop or workstation.

  • Safely dispose of tips, tubes, gloves and more
  • Secures and covers hazardous waste
  • Easy push open/close spring lid
  • Compact footprint (7×5×7")

For the temporary storage of biohazardous material during a lab activity or assay that doesn't require multiple trips to the biohazard bin. The sleek and compact design requires minimal space on the lab bench while the closed-lid system and easy bag removal creates a safe and convenient solution for waste .

Ordering information: Red biohazard bags (8.5×11") are purchased separately in packs of 100.

Delivery information: The BenchBin™ Starter Kit includes one BenchBin and 400 red 8.5×11" autoclave/ biohazard bags.

Spécifications
  • Désignation:
    BenchBin™ benchtop biohazard bin
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.