Test Tubes without Rim
175 ml, 38×200 mm
Catalog # 76532-236
Supplier: United Scientific Supplies
: TT9820-L
Jump to:
Product Details & Documents
Pre-cleaned borosilicate glass test tubes without rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
- Tubes do not have rims
- Autoclavable
Rimless borosilicate glass test tubes are ideal for a variety of laboratory applications.
Specifications
- Capacité:175 ml
- Diameter ×H:38×200 mm