Test Tubes without Rim
125 ml, 32×200 mm
Catalog # 76532-234
Supplier: United Scientific Supplies
Fournisseur #: TT9820-K
Jump to:
Product Details & Documents
Pre-cleaned borosilicate glass test tubes without rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
- Tubes do not have rims
- Autoclavable
Rimless borosilicate glass test tubes are ideal for a variety of laboratory applications.
Spécifications
- Capacité:125 ml
- Diameter ×H:32×200 mm