Test Tubes without Rim
37 ml, 25×100 mm
76532-228
: TT9820-H
Pre-cleaned borosilicate glass test tubes without rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
- Tubes do not have rims
- Autoclavable
Rimless borosilicate glass test tubes are ideal for a variety of laboratory applications.
- Capacité:37 ml
- Diameter ×H:25×100 mm