Test Tubes without Rim
Test Tubes without Rim
5 ml, 12×75 mm
Catalog # 76532-222
Supplier #:  TT9820-B
Test Tubes without Rim
Catalog # 76532-222
Test Tubes without Rim
Test Tubes without Rim
5 ml, 12×75 mm
Price
$37.79
New Customer Promotional Price
Log in for availability
Pack of 72
Product Details & Documents
Pre-cleaned borosilicate glass test tubes without rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
  • Tubes do not have rims
  • Autoclavable
Rimless borosilicate glass test tubes are ideal for a variety of laboratory applications.
Specifications
  • Capacité:
    5 ml
  • Diameter ×H:
    12×75 mm