Test Tubes without Rim
5 ml, 12×75 mm
Catalog # 76532-222
Supplier: United Scientific Supplies
Supplier #: TT9820-B
Jump to:
Product Details & Documents
Pre-cleaned borosilicate glass test tubes without rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
- Tubes do not have rims
- Autoclavable
Rimless borosilicate glass test tubes are ideal for a variety of laboratory applications.
Specifications
- Capacité:5 ml
- Diameter ×H:12×75 mm