Test Tubes with Rim
Test Tubes with Rim
16 ml, 16×125 mm
Catalog # 76532-216
Supplier #:  TT9800-M
Test Tubes with Rim
Catalog # 76532-216
Test Tubes with Rim
Test Tubes with Rim
16 ml, 16×125 mm
Price
$76.27
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 72
Product Details & Documents
Pre-cleaned borosilicate glass test tubes with rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
  • Tubes have rims
  • Autoclavable
Made from borosilicate glass, these general-purpose tubes will withstand daily use at an affordable price. Tubes have a uniform thickness and a strong lip.
Spécifications
  • Capacité:
    16 ml
  • Diameter ×H:
    16×125 mm