Vikan® Basic Wall Brackets, Remco
Yellow
Catalog # 76530-418
Supplier: Remco Products
Supplier #: 6156
Jump to:
Product Details & Documents
The wall bracket supports any product and is virtually unbreakable.
- Provides a color-coded storage option for tools
- Polypropylene construction
The one-piece bracket kit includes screws, wall anchors and mounting instructions.
Overall dimensions: 9.4L×6.5W×2.25"H
Specifications
- Color:Yellow