Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Vikan® Hygienic Wall Bracket, Double Hook Module, Remco | Avantor

Vikan® Hygienic Wall Bracket, Double Hook Module, Remco

Grey, 3.27×3.07×1.89"
Catalog # 76528-692
Supplier:   Remco Products
Fournisseur #:  101488
Vikan® Hygienic Wall Bracket, Double Hook Module, Remco
Catalog # 76528-692
Supplier:  Remco Products
Vikan® Hygienic Wall Bracket, Double Hook Module, Remco
Vikan® Hygienic Wall Bracket, Double Hook Module, Remco

Grey, 3.27×3.07×1.89"

Price
$28.81
Log in pour la disponibilité
Each
Product Details & Documents

Double hook module is designed for 1-2 cleaning tools that have a hanging hole with a D- or T- shapped handle.

  • Can hold products weighing up to 6.61 lbs.
  • Polypropylene/polyamide construction

The double hook module is easy to disassemble for cleaning or replacement.

Spécifications
  • Color:
    Grey
  • Dimensions:
    3.27×3.07×1.89"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.