Vikan® Hygienic Wall Bracket, Double Hook Module, Remco
Red, 3.27×3.07×1.89"
Catalog # 76528-678
Supplier: Remco Products
: 10144
Vikan® Hygienic Wall Bracket, Double Hook Module, Remco
Catalog # 76528-678
Supplier: Remco Products
Jump to:
Product Details & Documents
Double hook module is designed for 1-2 cleaning tools that have a hanging hole with a D- or T- shapped handle.
- Can hold products weighing up to 6.61 lbs.
- Polypropylene/polyamide construction
The double hook module is easy to disassemble for cleaning or replacement.
Spécifications
- Color:Red
- Dimensions:3.27×3.07×1.89"