Vikan® Hygienic Wall Bracket, Single Hook Module, Remco | Avantor
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Purple, 1.63×3.07×1.89"
Catalog # 76528-642
Supplier:   Remco Products
Fournisseur #:  10128
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Catalog # 76528-642
Supplier:  Remco Products
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Purple, 1.63×3.07×1.89"
Price
$21.66
Log in pour la disponibilité
Each
Product Details & Documents
Single hook module is designed for storing cleaning tools that have a hanging hole.
  • Can hold products weighing up to 6.61 lbs.
  • Polypropylene/polyamide construction
The hook system is easy to disassemble for cleaning or replacement.
Spécifications
  • Color:
    Purple
  • Dimensions:
    1.63×3.07×1.89"