Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Brown, 1.63×3.07×1.89"
76528-636
: 101266
Single hook module is designed for storing cleaning tools that have a hanging hole.
- Can hold products weighing up to 6.61 lbs.
- Polypropylene/polyamide construction
The hook system is easy to disassemble for cleaning or replacement.
- Color:Brown
- Dimensions:1.63×3.07×1.89"