Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
White, 1.63×3.07×1.89"
Catalog # 76528-632
Supplier: Remco Products
Fournisseur #: 10125
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Catalog # 76528-632
Supplier: Remco Products
Jump to:
Product Details & Documents
Single hook module is designed for storing cleaning tools that have a hanging hole.
- Can hold products weighing up to 6.61 lbs.
- Polypropylene/polyamide construction
The hook system is easy to disassemble for cleaning or replacement.
Spécifications
- Color:White
- Dimensions:1.63×3.07×1.89"